2-(Trimethylammonio)acetate 2-hydroxypropane-1,2,3-tricarboxylic acid salt ≥95%

Supplier: Ambeed
Warning

A111835-1000G A111835-25G A111835-500G A111835-5G A111835-100G
77592-194EA 51.25 USD
77592-194 77599-610 77599-612 77599-614 77623-926
2-(Trimethylammonio)acetate 2-hydroxypropane-1,2,3-tricarboxylic acid salt ≥95%
2-(Trimethylammonio)acetate 2-hydroxypropane-1,2,3-tricarboxylic acid salt

Precaución: for Research Use Only

Formula: C₁₁H₁₉NO₉
MDL Number: MFCD06411579
CAS Number: 17671-50-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR