2-(Chloromethyl)-3H-imidazo[4,5-b]pyridine hydrochloride ≥97%

Supplier: Ambeed
Danger

A1116597-250MG A1116597-100MG A1116597-5G
77595-748EA 102.5 USD
77595-748 77865-790 77865-791
2-(Chloromethyl)-3H-imidazo[4,5-b]pyridine hydrochloride ≥97%
2-(Chloromethyl)-3H-imidazo[4,5-b]pyridine hydrochloride

Precaución: for Research Use Only

Formula: C₇H₇Cl₂N₃
MW: 204.0566 g/mol
MDL Number: MFCD16622061
CAS Number: 24638-21-9
UN: 1759
ADR: 8 Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR