4-Methylpentan-1-amine hydrochloride ≥95%

Supplier: Ambeed
Warning

A1115014-1G A1115014-5G
77865-787EA 157 USD
77865-787 77865-788
4-Methylpentan-1-amine hydrochloride ≥95%
4-Methylpentan-1-amine hydrochloride

Precaución: for Research Use Only

Formula: C₆H₁₆ClN
MW: 137.65 g/mol
MDL Number: MFCD16040097
CAS Number: 116008-13-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR