2-Amino-N-(prop-2-yn-1-yl)acetamide hydrochloride ≥95%

Supplier: Ambeed
Warning

A1114580-1G A1114580-250MG
77595-742EA 98.4 USD
77595-742 77619-856
2-Amino-N-(prop-2-yn-1-yl)acetamide hydrochloride ≥95%
2-Amino-N-(prop-2-yn-1-yl)acetamide hydrochloride

Precaución: for Research Use Only

Formula: C₅H₉ClN₂O
MW: 148.5908 g/mol
MDL Number: MFCD15209627
CAS Number: 1221722-25-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR