2-Methyl-1,2,3,4-tetrahydroisoquinolin-5-amine ≥95%

Supplier: Ambeed
Warning

A1113810-1G A1113810-250MG
77570-640EA 380.07 USD
77570-640 77619-852
2-Methyl-1,2,3,4-tetrahydroisoquinolin-5-amine ≥95%
2-Methyl-1,2,3,4-tetrahydroisoquinolin-5-amine

Precaución: for Research Use Only

Formula: C₁₀H₁₄N₂
MW: 162.2316 g/mol
MDL Number: MFCD12778192
CAS Number: 14097-41-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR