Methyl (R)-4-(1-hydroxyethyl)benzoate ≥95%

Supplier: Ambeed
Warning

A1112495-1G A1112495-100MG
77619-850EA 407.88 USD
77619-850 77865-785
Methyl (R)-4-(1-hydroxyethyl)benzoate ≥95%
Methyl (R)-4-(1-hydroxyethyl)benzoate

Precaución: for Research Use Only

Formula: C₁₀H₁₂O₃
MDL Number: MFCD08702978
CAS Number: 129446-47-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR