3-Bromonaphthalen-1-amine ≥96%, Ambeed.Inc
Supplier: Ambeed
|
Warning
|
A111071-1G
A111071-250MG
77683-015EA
171
USD
77683-015
77891-798
3-Bromonaphthalen-1-amine ≥96%, Ambeed.Inc
3-Bromonaphthalen-1-amine
Precaución: For research use only.
Formula:
C₁₀H₈BrN |
MDL Number:
MFCD13180525 CAS Number: 90766-34-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...