2,2',4,4',6,6'-Hexamethyl-1,1'-biphenyl ≥95%

Supplier: Ambeed
Warning

A111025-100MG A111025-250MG
77885-020EA 36 USD
77885-020 77885-021
2,2',4,4',6,6'-Hexamethyl-1,1'-biphenyl ≥95%
2,2',4,4',6,6'-Hexamethyl-1,1'-biphenyl

Precaución: for Research Use Only

Formula: C₁₈H₂₂
MDL Number: MFCD00506060
CAS Number: 4482-03-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR