Ethyl 5-amino-2-methylnicotinate ≥97%

Supplier: Ambeed
Warning

A111014-100MG A111014-1G A111014-250MG
77885-017EA 33 USD
77885-017 77885-018 77885-019
Ethyl 5-amino-2-methylnicotinate ≥97%
Ethyl 5-amino-2-methylnicotinate

Precaución: for Research Use Only

Formula: C₉H₁₂N₂O₂
MDL Number: MFCD09909700
CAS Number: 60390-42-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR