tert-Butyl 6-(methylamino)-2-azaspiro[3.3]heptane-2-carboxylate ≥97%

Supplier: Ambeed
Warning

A110960-1G A110960-100MG A110960-250MG
77570-622EA 108.15 USD
77570-622 77747-819 77747-820
tert-Butyl 6-(methylamino)-2-azaspiro[3.3]heptane-2-carboxylate ≥97%
tert-Butyl 6-(methylamino)-2-azaspiro[3.3]heptane-2-carboxylate

Precaución: for Research Use Only

Formula: C₁₂H₂₂N₂O₂
MW: 226.32 g/mol
MDL Number: MFCD18642567
CAS Number: 1824024-00-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR