4-(Chlorosulfonyl)-2-hydroxybenzoic acid ≥95%

Supplier: Ambeed
Danger

A1107978-1G A1107978-100MG A1107978-250MG
77619-838EA 645.81 USD
77619-838 77783-211 77783-212
4-(Chlorosulfonyl)-2-hydroxybenzoic acid ≥95%
4-(Chlorosulfonyl)-2-hydroxybenzoic acid

Precaución: for Research Use Only

Formula: C₇H₅ClO₅S
MDL Number: MFCD10697740
CAS Number: 98273-15-5
UN: 3261
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR