2-Fluorobenzenesulfonamide ≥98%

Supplier: Ambeed
Danger

A110731-25G A110731-5G
77570-610EA 41 USD
77570-610 77747-818
2-Fluorobenzenesulfonamide ≥98%
2-Fluorobenzenesulphonamide

Precaución: for Research Use Only

Formula: C₆H₆FNO₂S
MDL Number: MFCD00042283
CAS Number: 30058-40-3
UN: 2811
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR