Chloromethyl 3-((tert-butoxycarbonyl)amino)propanoate ≥95%

Supplier: Ambeed
Danger

A1107245-100MG A1107245-250MG
77783-209EA 11 USD
77783-209 77783-210
Chloromethyl 3-((tert-butoxycarbonyl)amino)propanoate ≥95%
Chloromethyl 3-((tert-butoxycarbonyl)amino)propanoate

Precaución: for Research Use Only

Formula: C₉H₁₆ClNO₄
MDL Number: MFCD20268528
CAS Number: 150109-41-4
UN: 1760
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR