4-Oxo-4-((4-sulfamoylphenyl)amino)butanoic acid ≥98%

Supplier: Ambeed
Warning

A110709-250MG A110709-1G
77570-606EA 14.42 USD
77570-606 77666-620
4-Oxo-4-((4-sulfamoylphenyl)amino)butanoic acid ≥98%
4-Oxo-4-((4-sulfamoylphenyl)amino)butanoic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₂N₂O₅S
MW: 272.27 g/mol
MDL Number: MFCD00025390
CAS Number: 3563-14-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR