3-Bromo-2-fluoro-6-methyl-benzoic acid ≥95%
Supplier: Ambeed
|
Warning
|
Synonyms:
3-Bromo-2-fluoro-6-methylbenzoic acid
A1105968-1G
A1105968-100MG
A1105968-250MG
77570-594EA
492
USD
77570-594
77783-203
77783-204
3-Bromo-2-fluoro-6-methyl-benzoic acid ≥95%
3-Bromo-2-fluoro-6-methyl-benzoic acid
Precaución: for Research Use Only
Formula:
C₈H₆BrFO₂ |
MDL Number:
MFCD23712077 CAS Number: 1427433-22-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...