tert-Butyl 2-(2-((6-chlorohexyl)oxy)ethoxy)acetate ≥98%
Supplier: Ambeed
|
Warning
|
Synonyms:
Boc-C1-PEG2-C4-Cl
A110551-100MG
A110551-1G
A110551-250MG
A110551-50MG
77747-813EA
225
USD
77747-813
77747-814
77747-815
77747-816
tert-Butyl 2-(2-((6-chlorohexyl)oxy)ethoxy)acetate ≥98%
tert-Butyl 2-(2-((6-chlorohexyl)oxy)ethoxy)acetate
Precaución: for Research Use Only
Formula:
C₁₄H₂₇ClO₄ |
MDL Number:
MFCD31692348 CAS Number: 1835705-53-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...