Aclidinium bromide ≥98%

Supplier: Ambeed
Warning

A110489-1MG A110489-5MG A110489-10MG A110489-50MG
77599-588EA 6.15 USD
77599-588 77599-590 77623-914 77876-824
Aclidinium bromide ≥98%
Aclidinium bromide

Precaución: for Research Use Only

Formula: C₂₆H₃₀NO₄S₂·Br
MDL Number: MFCD09954105
CAS Number: 320345-99-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR