4,7-Dihydroxy-3-(4-methoxyphenyl)-2H-chromen-2-one ≥95%

Supplier: Ambeed
Warning

A1104840-100MG A1104840-250MG
77783-195EA 16 USD
77783-195 77783-196
4,7-Dihydroxy-3-(4-methoxyphenyl)-2H-chromen-2-one ≥95%
4,7-Dihydroxy-3-(4-methoxyphenyl)-2H-chromen-2-one

Precaución: for Research Use Only

Formula: C₁₆H₁₂O₅
MDL Number: MFCD08548200
CAS Number: 42345-56-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR