3-(4-Iodophenyl)-3-(trifluoromethyl)-3H-diazirine ≥97%

Supplier: Ambeed
Danger

A110477-100MG A110477-250MG
77614-852EA 204.97 USD
77614-852 77747-812
3-(4-Iodophenyl)-3-(trifluoromethyl)-3H-diazirine ≥97%
3-(4-Iodophenyl)-3-(trifluoromethyl)-3H-diazirine

Precaución: for Research Use Only

Formula: C₈H₄F₃IN₂
MW: 312.03 g/mol
MDL Number: MFCD25960903
CAS Number: 210107-38-3
UN: 2924
ADR: 3,8,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR