PXD101 ≥97%

Supplier: Ambeed
Warning

A110435-1G A110435-250MG
77666-614EA 118 USD
77666-614 77666-616
PXD101 ≥97%
PXD101

Precaución: for Research Use Only

Formula: C₁₅H₁₄N₂O₄S
MDL Number: MFCD08064035
CAS Number: 414864-00-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR