2-Chlorothiophene-3-carboxaldehyde ≥98%

Supplier: Ambeed
Warning

A110096-1G A110096-5G
77610-872EA 30.9 USD
77610-872 77889-761
2-Chlorothiophene-3-carboxaldehyde ≥98%
2-Chlorothiophene-3-carboxaldehyde

Precaución: for Research Use Only

Formula: C₅H₃ClOS
MDL Number: MFCD08694817
CAS Number: 14345-98-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR