4-Iodo-1-(tetrahydro-2H-pyran-2-yl)-1H-pyrazole ≥98%

Supplier: Ambeed
Warning

A109827-5G A109827-1G A109827-25G
77610-870EA 77.25 USD
77610-870 77670-554 77880-890
4-Iodo-1-(tetrahydro-2H-pyran-2-yl)-1H-pyrazole ≥98%
4-Iodo-1-(tetrahydro-2H-pyran-2-yl)-1H-pyrazole

Precaución: for Research Use Only

Formula: C₈H₁₁IN₂O
MDL Number: MFCD19689194
CAS Number: 938066-17-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR