3-(Ethylamino)benzonitrile ≥95%

Supplier: Ambeed
Danger

A1095598-1G A1095598-250MG
77783-186EA 109 USD
77783-186 77783-187
3-(Ethylamino)benzonitrile ≥95%
3-(Ethylamino)benzonitrile

Precaución: for Research Use Only

Formula: C₉H₁₀N₂
MDL Number: MFCD11144774
CAS Number: 883742-00-5
UN: 3276
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR