3,3'-Diethyloxacarbocyanine iodide [DiOC2(3) iodide] ≥98%

Supplier: Ambeed
Warning

Synonyms: DOC Iodide, 3-Ethyl-2-[3-[3-ethyl-2(3H)-benzoxazolylidene]-1-propenyl]benzoxazolium Iodide, 3,3'-Diethyloxacarbocyanine iodide, DiOC2(3) iodide, 3-Ethyl-2-(3-(3-ethylbenzo[d]oxazol-2(3H)-ylidene)prop-1-en-1-yl)benzo[d]oxazol-3-ium iodide

A109271-1G A109271-250MG A109271-5G
77610-864EA 79.31 USD
77610-864 77684-983 77684-984
3,3'-Diethyloxacarbocyanine iodide [DiOC2(3) iodide] ≥98%
3,3'-Diethyloxacarbocyanine iodide [DiOC2(3) iodide]

Precaución: for Research Use Only

Formula: C₂₁H₂₁IN₂O₂
MW: 460.32 g/mol
MDL Number: MFCD00011952
CAS Number: 905-96-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR