1-Boc-4-cyano-4-methylpiperidine ≥98%

Supplier: Ambeed
Danger

Synonyms: tert-Butyl 4-cyano-4-methylpiperidine-1-carboxylate

A109128-1G A109128-250MG
77882-914EA 39 USD
77882-914 77882-915
1-Boc-4-cyano-4-methylpiperidine ≥98%
1-Boc-4-cyano-4-methylpiperidine

Precaución: for Research Use Only

Formula: C₁₂H₂₀N₂O₂
MDL Number: MFCD11849789
CAS Number: 530115-96-9
UN: 2811
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR