3-(Aminomethyl)-5-chloroaniline ≥92%

Supplier: Ambeed
Danger

A1090473-250MG A1090473-1G
77619-820EA 123.6 USD
77619-820 77783-175
3-(Aminomethyl)-5-chloroaniline ≥92%
3-(Aminomethyl)-5-chloroaniline

Precaución: for Research Use Only

Formula: C₇H₉ClN₂
MDL Number: MFCD08705641
CAS Number: 683740-35-4
UN: 3259
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR