3-Bromo-2-(trifluoromethoxy)pyridine ≥97%

Supplier: Ambeed
Warning

A109042-100MG A109042-250MG
77901-957EA 296 USD
77901-957 77900-807
3-Bromo-2-(trifluoromethoxy)pyridine ≥97%
3-Bromo-2-(trifluoromethoxy)pyridine

Precaución: for Research Use Only

Formula: C₆H₃BrF₃NO
MDL Number: MFCD11506285
CAS Number: 1086393-00-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR