N-Boc-6-hydroxy-3-azabicyclo[3.2.0]heptane ≥98%

Supplier: Ambeed
Warning

A108966-100MG A108966-1G
77871-432EA 49 USD
77871-432 77871-433
N-Boc-6-hydroxy-3-azabicyclo[3.2.0]heptane ≥98%
N-Boc-6-hydroxy-3-azabicyclo[3.2.0]heptane

Precaución: for Research Use Only

Formula: C₁₁H₁₉NO₃
MDL Number: MFCD12031304
CAS Number: 663172-78-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR