5-Bromo-1,3-oxazole hydrochloride ≥95%

Supplier: Ambeed

A1089574-1G A1089574-250MG
77570-518EA 370 USD
77570-518 77783-172
5-Bromo-1,3-oxazole hydrochloride ≥95%
5-Bromo-1,3-oxazole hydrochloride

Precaución: for Research Use Only

Formula: C₃H₃BrClNO
MDL Number: MFCD29924285
CAS Number: 1955557-64-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR