1-(6-(4-Fluorobenzyl)-5-(hydroxymethyl)-3,3-dimethyl-2,3-dihydro-1H-pyrrolo[3,2-b]pyridin-1-yl)-2-((2R,5R)-5-methyl-2-(((R)-3-methylmorpholino)methyl)piperazin-1-yl)ethanone 98+%

Supplier: Ambeed
Warning

Synonyms: Tolinapant

A108877-10MG A108877-50MG
77614-832EA 229.69 USD
77614-832 77747-804
1-(6-(4-Fluorobenzyl)-5-(hydroxymethyl)-3,3-dimethyl-2,3-dihydro-1H-pyrrolo[3,2-b]pyridin-1-yl)-2-((2R,5R)-5-methyl-2-(((R)-3-methylmorpholino)methyl)piperazin-1-yl)ethanone 98+%
1-(6-(4-Fluorobenzyl)-5-(hydroxymethyl)-3,3-dimethyl-2,3-dihydro-1H-pyrrolo[3,2-b]pyridin-1-yl)-2-((2R,5R)-5-methyl-2-(((R)-3-methylmorpholino)methyl)piperazin-1-yl)ethanone

Precaución: for Research Use Only

Formula: C₃₀H₄₂FN₅O₃
MW: 539.68 g/mol
MDL Number: MFCD31619274
CAS Number: 1799328-86-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR