Ethyl 2-((ethoxycarbonyl)amino)-4-methyl-5-(4-nitrophenyl)thiophene-3-carboxylate ≥97%

Supplier: Ambeed
Warning

A108855-1G A108855-100G A108855-25G A108855-5G
77614-830EA 14.42 USD
77614-830 77747-801 77747-802 77747-803
Ethyl 2-((ethoxycarbonyl)amino)-4-methyl-5-(4-nitrophenyl)thiophene-3-carboxylate ≥97%
Ethyl 2-((ethoxycarbonyl)amino)-4-methyl-5-(4-nitrophenyl)thiophene-3-carboxylate

Precaución: for Research Use Only

Formula: C₁₇H₁₈N₂O₆S
MDL Number: MFCD31811620
CAS Number: 308831-93-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR