(3R,4S,5S)-tert-Butyl 3-methoxy-5-methyl-4-(methylamino)heptanoate hydrochloride ≥97%
Supplier: Ambeed
|
Warning
|
Synonyms:
Monomethyl auristatin E intermediate-1
A108652-250MG
A108652-1G
A108652-5G
77592-170EA
58.43
USD
77592-170
77682-997
77682-998
(3R,4S,5S)-tert-Butyl 3-methoxy-5-methyl-4-(methylamino)heptanoate hydrochloride ≥97%
(3R,4S,5S)-tert-Butyl 3-methoxy-5-methyl-4-(methylamino)heptanoate hydrochloride
Precaución: For research use only.
Formula:
C₁₄H₃₀ClNO₃ MW: 295.85 g/mol |
MDL Number:
MFCD19688575 CAS Number: 120205-48-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...