2-Cyano-4-methylbenzene-1-sulfonyl chloride ≥98%

Supplier: Ambeed
Danger

A1086395-100MG A1086395-250MG
77619-816EA 118.45 USD
77619-816 77783-161
2-Cyano-4-methylbenzene-1-sulfonyl chloride ≥98%
2-Cyano-4-methylbenzene-1-sulfonyl chloride

Precaución: for Research Use Only

Formula: C₈H₆ClNO₂S
MDL Number: MFCD18394317
CAS Number: 1261682-98-7
UN: 3261
ADR: 8,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR