(R)-1-(4-(Trifluoromethoxy)phenyl)ethanamine hydrochloride ≥95%

Supplier: Ambeed
Warning

A108600-100MG A108600-250MG
77891-791EA 46 USD
77891-791 77891-792
(R)-1-(4-(Trifluoromethoxy)phenyl)ethanamine hydrochloride ≥95%
(R)-1-(4-(Trifluoromethoxy)phenyl)ethanamine hydrochloride

Precaución: for Research Use Only

Formula: C₉H₁₁ClF₃NO
MDL Number: MFCD12757221
CAS Number: 1208989-29-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR