2-Acetylthiazole-5-carboxylic acid ≥98%

Supplier: Ambeed
Warning

A108567-100MG A108567-250MG
77882-911EA 60 USD
77882-911 77882-912
2-Acetylthiazole-5-carboxylic acid ≥98%
2-Acetylthiazole-5-carboxylic acid

Precaución: for Research Use Only

Formula: C₆H₅NO₃S
MDL Number: MFCD17169900
CAS Number: 1095824-76-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR