Methyl 3-iodo-1-methyl-1H-indazole-5-carboxylate 98+%

Supplier: Ambeed
Warning

A108497-5G A108497-1G
77684-975EA 56 USD
77684-975 77891-790
Methyl 3-iodo-1-methyl-1H-indazole-5-carboxylate 98+%
Methyl 3-iodo-1-methyl-1H-indazole-5-carboxylate

Precaución: for Research Use Only

Formula: C₁₀H₉IN₂O₂
MDL Number: MFCD15071440
CAS Number: 1234616-44-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR