3-(Oxolan-3-yl)-1H-pyrazol-5-amine ≥95%

Supplier: Ambeed
Warning

A1082913-250MG A1082913-100MG
77619-808EA 206 USD
77619-808 77783-145
3-(Oxolan-3-yl)-1H-pyrazol-5-amine ≥95%
3-(Oxolan-3-yl)-1H-pyrazol-5-amine

Precaución: for Research Use Only

Formula: C₇H₁₁N₃O
MDL Number: MFCD21647355
CAS Number: 1186609-16-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR