3-Amino-1-methylpiperidine-2,6-dione hydrochloride ≥98%

Supplier: Ambeed

A1081961-100MG A1081961-1G A1081961-250MG A1081961-5G
77783-138EA 136 USD
77783-138 77783-139 77783-140 77783-141
3-Amino-1-methylpiperidine-2,6-dione hydrochloride ≥98%
3-Amino-1-methylpiperidine-2,6-dione hydrochloride

Precaución: for Research Use Only

Formula: C₆H₁₁ClN₂O₂
MW: 178.62 g/mol
MDL Number: MFCD28895640
CAS Number: 1909304-98-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR