3-sulfopropanoic acid ≥95%

Supplier: Ambeed
Danger

A1081388-100MG A1081388-50MG
77783-136EA 150 USD
77783-136 77783-137
3-sulfopropanoic acid ≥95%
3-sulfopropanoic acid

Precaución: for Research Use Only

Formula: C₃H₆O₅S
MDL Number: MFCD19347139
CAS Number: 44826-45-1
UN: 3261
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR