N-[4-(1-Benzofuran-2-yl)-1,3-thiazol-2-yl]-2-chloroacetamide ≥95%

Supplier: Ambeed
Danger

A1080283-1G A1080283-250MG
77783-122EA 36 USD
77783-122 77783-123
N-[4-(1-Benzofuran-2-yl)-1,3-thiazol-2-yl]-2-chloroacetamide ≥95%
N-[4-(1-Benzofuran-2-yl)-1,3-thiazol-2-yl]-2-chloroacetamide

Precaución: for Research Use Only

Formula: C₁₃H₉ClN₂O₂S
MDL Number: MFCD08444302
CAS Number: 924129-01-1
UN: 1759
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR