2-Chloro-N-(2-cyanoethyl)-N-(3,4-dimethylphenyl)acetamide ≥95%

Supplier: Ambeed

A1080226-1G A1080226-250MG
77783-120EA 684 USD
77783-120 77783-121
2-Chloro-N-(2-cyanoethyl)-N-(3,4-dimethylphenyl)acetamide ≥95%
2-Chloro-N-(2-cyanoethyl)-N-(3,4-dimethylphenyl)acetamide

Precaución: for Research Use Only

Formula: C₁₃H₁₅ClN₂O
MDL Number: MFCD00700886
CAS Number: 324009-92-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR