Etrasimod ≥98%

Supplier: Ambeed
Warning

Synonyms: APD-334, APD334, (R)-2-(7-((4-Cyclopentyl-3-(trifluoromethyl)benzyl)oxy)-1,2,3,4-tetrahydrocyclopenta[b]indol-3-yl)acetic acid, (3R)-7-[[4-cyclopentyl-3-(trifluoromethyl)phenyl]methoxy]-1,2,3,4-tetrahydro-cyclopent[b]indole-3-acetic acid

A107759-10MG A107759-1MG A107759-5MG
77614-816EA 87.55 USD
77614-816 77614-818 77614-820
Etrasimod ≥98%
Etrasimod

Precaución: for Research Use Only

Formula: C₂₆H₂₆F₃NO₃
MDL Number: MFCD28502134
CAS Number: 1206123-37-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR