Ethyl 3-aminoquinoline-2-carboxylate ≥95%

Supplier: Ambeed
Warning

A107507-250MG A107507-5G
77610-848EA 46.35 USD
77610-848 77670-536
Ethyl 3-aminoquinoline-2-carboxylate ≥95%
Ethyl 3-aminoquinoline-2-carboxylate

Precaución: for Research Use Only

Formula: C₁₂H₁₂N₂O₂
MDL Number: MFCD18427520
CAS Number: 62235-59-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR