Genistein ≥98%

Supplier: Ambeed
Warning

A107459-1G A107459-10G A107459-5G
77599-558EA 7.18 USD
77599-558 77614-806 77614-808
Genistein ≥98%
Genistein

Precaución: for Research Use Only

Formula: C₁₅H₁₀O₅
MDL Number: MFCD00016952
CAS Number: 446-72-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR