4-Chloro-benzenesulfonyl fluoride ≥95%

Supplier: Ambeed
Danger

A1072605-250MG A1072605-1G A1072605-25G
77619-790EA 13.39 USD
77619-790 77783-108 77783-109
4-Chloro-benzenesulfonyl fluoride ≥95%
4-Chloro-benzenesulfonyl fluoride

Precaución: for Research Use Only

Formula: C₆H₄ClFO₂S
MDL Number: MFCD01657965
CAS Number: 349-89-3
UN: 3261
ADR: 8,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR