3-Bromo-2,4-dimethoxypyridine 95+%

Supplier: Ambeed
Warning

A106912-1G A106912-250MG
77884-974EA 125 USD
77884-974 77884-975
3-Bromo-2,4-dimethoxypyridine 95+%
3-Bromo-2,4-dimethoxypyridine

Precaución: for Research Use Only

Formula: C₇H₈BrNO₂
MDL Number: MFCD19441222
CAS Number: 96246-00-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR