3-Ethenyl-1H-indole ≥95% stabilized

Supplier: Ambeed
Warning


Stabilization: Stabilised with TBC

A1067350-1G A1067350-250MG
77783-076EA 449 USD
77783-076 77783-077
3-Ethenyl-1H-indole ≥95% stabilized
3-Ethenyl-1H-indole

Precaución: for Research Use Only

Formula: C₁₀H₉N
MDL Number: MFCD19217205
CAS Number: 53654-36-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR