tert-Butyl 3-sulfanylpropanoate ≥95%

Supplier: Ambeed
Danger

A1067052-1G A1067052-100MG
77570-400EA 343 USD
77570-400 77783-075
tert-Butyl 3-sulfanylpropanoate ≥95%
tert-Butyl 3-sulfanylpropanoate

Precaución: for Research Use Only

Formula: C₇H₁₄O₂S
MDL Number: MFCD28246452
CAS Number: 59854-14-7
UN: 2810
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR