N-(2-Methoxy-5-(4,4,5,5-Tetramethyl-1,3,2-Dioxaborolan-2-Yl)Pyridin-3-Yl)Methanesulfonamide ≥95%, Ambeed.Inc

Supplier: Ambeed
Warning

A106539-250MG A106539-100MG A106539-1G
77592-138EA 133.25 USD
77592-138 77610-842 77682-980
N-(2-Methoxy-5-(4,4,5,5-Tetramethyl-1,3,2-Dioxaborolan-2-Yl)Pyridin-3-Yl)Methanesulfonamide ≥95%, Ambeed.Inc
N-(2-Methoxy-5-(4,4,5,5-Tetramethyl-1,3,2-Dioxaborolan-2-Yl)Pyridin-3-Yl)Methanesulfonamide

Precaución: For research use only.

Formula: C₁₃H₂₁BN₂O₅S
MDL Number: MFCD12964555
CAS Number: 1083326-75-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR