4-Fluoro-2,6-dimethylbenzene-1-sulfonyl chloride ≥95%

Supplier: Ambeed
Danger

A1064918-100MG A1064918-1G
77783-059EA 83 USD
77783-059 77783-060
4-Fluoro-2,6-dimethylbenzene-1-sulfonyl chloride ≥95%
4-Fluoro-2,6-dimethylbenzene-1-sulfonyl chloride

Precaución: for Research Use Only

Formula: C₈H₈ClFO₂S
MDL Number: MFCD19200538
CAS Number: 88972-03-6
UN: 3261
ADR: 8,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR